Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171739.1 | NP_001165210.1 | 1 | 469 | ||
NM_009736.3 | NP_033866.4 | 1 | 469 | |||
GenBank mRNA | AF022223.1 | - | 1 | 410 | ||
AK009149.1 | - | 1 | 380 | |||
AK154469.1 | - | 1 | 419 | |||
AK171094.1 | - | 1 | 419 | |||
BC003722.1 | - | 1 | 431 | 3495306 | ||
BC069918.1 | - | 1 | 406 | 4979705 | ||
BC093509.1 | - | 1 | 380 | 4978330 | ||
CA321856.1 | - | 1 | 456 | |||
CX225744.1 | - | 1 | 469 |