Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bub3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009774.3 | NP_033904.2 | 5, 6 | 648 | ||
GenBank mRNA | AF149822.1 | - | 4, 5 | 584 | ||
AK088534.1 | - | 5, 6 | 649 | |||
AK198565.1 | - | 1, 2 | 68 | |||
AK204610.1 | - | 1, 2 | 68 | |||
BC025089.1 | - | 5, 6 | 660 | 3586753 | ||
BC086790.1 | - | 3, 4 | 2613 | |||
U67327.1 | - | 5, 6 | 635 |