Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110794.1 | NP_001104264.1 | 13 | 1663 | ||
NM_009674.3 | NP_033804.2 | 13 | 1778 | |||
GenBank mRNA | AK077579.1 | - | 13 | 1471 | ||
AK134589.1 | - | 14 | 1548 | |||
AK149654.1 | - | 13 | 1469 | |||
AK151438.1 | - | 13 | 1472 | |||
AK161613.1 | - | 13 | 1472 | |||
AK167588.1 | - | 13 | 1446 | |||
AK169199.1 | - | 13 | 1445 | |||
AK178726.1 | - | 2 | 250 | |||
BC008997.1 | - | 13 | 1462 | 3157926 | ||
L13129.1 | - | 13 | 1465 |