Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110140.3 | NP_001103610.1 | 5 | 908 | ||
NM_009722.3 | NP_033852.1 | 5 | 908 | |||
GenBank mRNA | AF039893.1 | - | 1 | 55 | ||
AJ131821.1 | - | 5 | 578 | |||
AJ223584.1 | - | 5 | 578 | |||
AK220538.1 | - | 5 | 857 | |||
BC054531.1 | - | 5 | 567 | 6406916 | ||
BC054748.1 | - | 5 | 567 | 6406916 |