Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001036684.2 | NP_001031761.1 | 12 | 2587 | ||
NM_009723.5 | NP_033853.1 | 14 | 2776 | |||
GenBank mRNA | AB030737.1 | - | 11 | 2148 | ||
AB030738.1 | - | 11 | 2148 | |||
AF053471.1 | - | 13 | 2605 | |||
AK054112.1 | - | 16 | 2874 | |||
AK147240.1 | - | 14 | 2662 | |||
AK147357.1 | - | 12 | 2516 | |||
AK147369.1 | - | 14 | 2711 | |||
AK147399.1 | - | 12 | 2739 | |||
AK147501.1 | - | 14 | 2723 | |||
GU734816.2 | - | 14 | 2283 |