Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bub3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009774.3 | NP_033904.2 | 6 | 746 | ||
GenBank mRNA | AF149822.1 | - | 5 | 682 | ||
AK088534.1 | - | 6 | 747 | |||
AK186549.1 | - | 1 | 103 | |||
AK198565.1 | - | 2 | 166 | |||
AK205773.1 | - | 1 | 101 | |||
AK207337.1 | - | 1 | 101 | |||
AK210147.1 | - | 1 | 101 | |||
AK212486.1 | - | 1 | 101 | |||
BC025089.1 | - | 6 | 758 | 3586753 | ||
BC086790.1 | - | 4 | 2711 | |||
U67327.1 | - | 6 | 733 |