Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009788.4 | NP_033918.1 | 5, 6 | 506 | ||
GenBank mRNA | AK002635.1 | - | 5, 6 | 508 | ||
AK005081.1 | - | 5, 6 | 505 | |||
AK005243.1 | - | 5, 6 | 506 | |||
AK028050.1 | - | 5, 6 | 507 | |||
AK028327.1 | - | 5, 6 | 505 | |||
AK038856.1 | - | 507 | ||||
AK048517.1 | - | 5, 6 | 507 | |||
AK077499.1 | - | 5, 6 | 505 | |||
AK078358.1 | - | 5, 6 | 504 | |||
BC016421.1 | - | 5, 6 | 437 | 4527000 | ||
M21531.1 | - | 5, 6 | 486 | |||
M23663.1 | - | 2, 3 | 68 |