Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009804.2 | NP_033934.2 | 2 | 282 | ||
GenBank mRNA | AK075853.1 | - | 2 | 252 | ||
AK088168.1 | - | 2 | 244 | |||
AK128919.1 | - | 2 | 257 | |||
AK133881.1 | - | 2 | 250 | |||
AK148961.1 | - | 2 | 250 | |||
AK150893.1 | - | 2 | 253 | |||
AK159152.1 | - | 2 | 253 | |||
AK159885.1 | - | 2 | 253 | |||
AK159891.1 | - | 2 | 204 | |||
AK159913.1 | - | 2 | 234 | |||
AK169069.1 | - | 2 | 247 | |||
AY040626.1 | - | 2 | 223 | |||
BC013447.1 | - | 2 | 239 | 4241333 | ||
BC047126.1 | - | 2 | 197 | |||
BY093963.1 | - | 2 | 282 | |||
L25069.1 | - | 2 | 254 | |||
M62897.1 | - | 2 | 167 | |||
X52108.1 | - | 2 | 167 |