Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Runx1t1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111026.2 | NP_001104496.1 | 2 | 538 | ||
NM_001111027.2 | NP_001104497.1 | 3 | 732 | |||
NM_009822.3 | NP_033952.1 | 2 | 470 | |||
GenBank mRNA | AB221624.1 | - | 1 | 117 | ||
AK032132.1 | - | 2 | 405 | |||
AK049615.1 | - | 2 | 276 | |||
AK142006.1 | - | 2 | 539 | |||
BC139200.1 | - | 2 | 196 | 8862222 | ||
BY125464.1 | - | 2 | 203 | |||
D32007.1 | - | 2 | 470 | |||
X79989.1 | - | 2 | 177 |