Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009881.3 | NP_034011.1 | 4, 5 | 445 | ||
GenBank mRNA | AF081260.1 | - | 4, 5 | 427 | ||
AF081261.1 | - | 4, 5 | 427 | |||
AK028162.1 | - | 1, 2 | 191 | |||
BC055103.1 | - | 4, 5 | 394 | 6741468 |