Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042634.2 | NP_001036099.1 | 2 | 311 | ||
GenBank mRNA | AK031218.1 | - | 2 | 245 | ||
AK076199.1 | - | 2 | 246 | |||
AK135765.1 | - | 2 | 246 | |||
BC062887.1 | - | 263 | ||||
BC145905.1 | - | 2 | 214 | |||
BC145911.1 | - | 2 | 214 | |||
BY784955.1 | - | 2 | 312 | |||
L29221.1 | - | 2 | 227 | |||
M38381.1 | - | 2 | 227 | |||
U11054.1 | - | 2 | 211 | |||
U21209.1 | - | 2 | 227 | |||
X57186.1 | - | 2 | 211 |