Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009793.3 | NP_033923.2 | 4, 5 | 721 | ||
GenBank mRNA | AK030219.1 | - | 4, 5 | 605 | ||
AK043114.1 | - | 4, 5 | 431 | |||
BC070420.1 | - | 4, 5 | 602 | 5718423 | ||
X58995.1 | - | 4, 5 | 411 |