Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccne2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037134.2 | NP_001032211.1 | 5 | 418 | ||
NM_001282943.1 | NP_001269872.1 | 5 | 331 | |||
NM_009830.3 | NP_033960.1 | 5 | 328 | |||
GenBank mRNA | AF091432.1 | - | 4 | 223 | ||
AF106691.1 | - | 3 | 198 | |||
AK010929.1 | - | 5 | 258 | |||
AK031116.1 | - | 5 | 255 | |||
AK037791.1 | - | 5 | 258 | |||
AK158720.1 | - | 5 | 844 | |||
AK167428.1 | - | 5 | 258 | |||
BC053727.1 | - | 4 | 218 | 30061057 |