Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capzb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037761.2 | NP_001032850.1 | 7, 8 | 789 | ||
NM_001271405.1 | NP_001258334.1 | 7, 8 | 768 | |||
NM_001271406.1 | NP_001258335.1 | 7, 8 | 754 | |||
NM_009798.4 | NP_033928.1 | 7, 8 | 789 | |||
GenBank mRNA | AK153367.1 | - | 7, 8 | 566 | ||
AK153394.1 | - | 7, 8 | 566 | |||
AK156778.1 | - | 7, 8 | 758 | |||
AK159390.1 | - | 7, 8 | 763 | |||
AK160082.1 | - | 7, 8 | 764 | |||
AK162757.1 | - | 7, 8 | 756 | |||
AK168738.1 | - | 7, 8 | 717 | |||
AK177260.1 | - | 2, 3 | 117 | |||
AK177711.1 | - | 2, 3 | 116 | |||
AK182951.1 | - | 2, 3 | 118 | |||
AK186717.1 | - | 2, 3 | 117 | |||
AK187793.1 | - | 2, 3 | 117 | |||
AK188337.1 | - | 2, 3 | 117 | |||
AK190028.1 | - | 2, 3 | 118 | |||
AK192076.1 | - | 2, 3 | 117 | |||
AK207588.1 | - | 2, 3 | 117 | |||
AK212957.1 | - | 2, 3 | 180 | |||
AK219201.1 | - | 2, 3 | 117 | |||
BC002053.1 | - | 7, 8 | 731 | 3590980 | ||
FJ692320.1 | - | 7, 8 | 754 | |||
U10406.1 | - | 7, 8 | 709 | |||
U10407.1 | - | 7, 8 | 680 |