Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166625.1 | NP_001160097.1 | 4 | 557 | ||
NM_009913.6 | NP_034043.1 | 3 | 489 | |||
GenBank mRNA | AJ131357.2 | - | 4 | 494 | ||
AJ132336.1 | - | 3 | 455 | |||
AK019478.1 | - | 663 | ||||
AK050615.1 | - | 3 | 490 | |||
AK153975.1 | - | 3 | 490 | |||
BC132166.1 | - | 2 | 398 | 40130443 | ||
BC132466.1 | - | 2 | 398 | 40130743 |
Set Membership: |
G-protein coupled receptor |