Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF053770.1 | - | 3 | 422 | ||
AF080580.1 | - | 4 | 485 | |||
AF098949.1 | - | 4 | 551 | |||
AK137841.1 | - | 2 | 2516 | |||
AK158380.1 | - | 4 | 536 | |||
BC028276.1 | - | 2 | 227 | |||
BC038681.1 | - | 4 | 518 | 1380996 | ||
U81277.1 | - | 3 | 267 |