Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009909.3 | NP_034039.1 | 2 | 531 | ||
GenBank mRNA | AK158234.1 | - | 3 | 537 | ||
BC051677.1 | - | 3 | 504 | 30023687 | ||
CJ056061.1 | - | 2 | 532 | |||
D17630.1 | - | 2 | 527 |