Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006902.4 | NP_008833.1 | 5 | 823 | ||
NM_022716.3 | NP_073207.1 | 4 | 751 | |||
XM_006711388.3 | XP_006711451.1 | 5 | 814 | |||
GenBank mRNA | AB451463.1 | - | 4 | 704 | ||
AK225968.1 | - | 5 | 1830 | |||
BC074993.2 | - | 4 | 751 | 30915308 | ||
BI039001.1 | - | 2 | 237 |