Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col18a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109991.1 | NP_001103461.1 | 4, 5 | 1441 | ||
NM_009929.3 | NP_034059.2 | 5, 6 | 857 | |||
GenBank mRNA | AK031798.1 | - | 5, 6 | 845 | ||
BC043697.1 | - | 4, 5 | 1441 | |||
BC062931.1 | - | 5, 6 | 823 | |||
BC064817.1 | - | 5, 6 | 828 | 6332464 | ||
BC066080.1 | - | 5, 6 | 823 | 30355615 | ||
D17546.1 | - | 3, 4 | 251 | |||
L16898.1 | - | 5, 6 | 796 | |||
L22545.1 | - | 3, 4 | 695 |