Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010071.2 | NP_034201.1 | 4 | 518 | ||
GenBank mRNA | AF030627.1 | - | 4 | 518 | ||
AF035117.1 | - | 4 | 514 | |||
AF059583.1 | - | 4 | 504 | |||
AK155579.1 | - | 4 | 515 | |||
AK158246.1 | - | 4 | 482 | |||
AK159466.1 | - | 4 | 559 | |||
AK159472.1 | - | 4 | 596 | |||
BC004590.1 | - | 4 | 510 | 3587682 |