Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S1pr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010102.2 | NP_034232.1 | 1 | 1586 | ||
GenBank mRNA | AJ006074.1 | - | 1 | 1586 | ||
AK075854.1 | - | 1 | 1105 | |||
AK154242.1 | - | 1 | 1109 | |||
BC107000.1 | - | 1 | 1156 | 40045304 |
Set Membership: |
![]() |