Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054021.1 | NP_473362.1 | 1 | 200 | ||
GenBank mRNA | BC069439.1 | - | 1 | 200 | 7262356 |
Set Membership: |
G-protein coupled receptor |