Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgrt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010189.3 | NP_034319.2 | 4 | 809 | ||
GenBank mRNA | AK146933.1 | - | 4 | 774 | ||
AK152879.1 | - | 4 | 745 | |||
BC003786.1 | - | 4 | 744 | 3485649 | ||
D37874.1 | - | 4 | 742 | |||
L17022.1 | - | 3 | 448 |