Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289533.1 | NP_001276462.1 | 6 | 1368 | ||
NM_010212.4 | NP_034342.1 | 6 | 1517 | |||
GenBank mRNA | AF153340.1 | - | 5 | 1284 | ||
AK133708.1 | - | 6 | 1487 | |||
AK147024.1 | - | 6 | 1518 | |||
U77040.2 | - | 6 | 1407 |