Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010260.1 | NP_034390.1 | 11 | 2273 | ||
GenBank mRNA | AF077007.1 | - | 11 | 2273 | ||
AF109168.1 | - | 11 | 2226 | |||
AJ007970.1 | - | 11 | 2266 | |||
BC011336.1 | - | 2294 | 3494311 | |||
BC032882.1 | - | 11 | 2288 | 1230883 | ||
BC049141.1 | - | 11 | 2282 |