Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010282.2 | NP_034412.1 | 3 | 305 | ||
GenBank mRNA | AB016044.1 | - | 3 | 193 | ||
AK011980.1 | - | 4 | 450 | |||
AK132907.1 | - | 3 | 298 | |||
BC006798.1 | - | 3 | 290 | 2651229 | ||
BC069913.1 | - | 3 | 305 | 6469191 |