Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnaz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010311.3 | NP_034441.1 | 3 | 1160 | ||
GenBank mRNA | AF056972.1 | - | 1 | 95 | ||
AK080716.1 | - | 3 | 1160 | |||
AK133483.1 | - | 1 | 1036 | |||
AK134710.1 | - | 1 | 864 | |||
BC014702.1 | - | 1 | 861 | 4501587 |