Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pfdn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185182.1 | NP_001172111.1 | 5 | 477 | ||
NM_010385.2 | NP_034515.1 | 4 | 681 | |||
GenBank mRNA | AK010679.1 | - | 4 | 671 | ||
AK076015.1 | - | 4 | 664 | |||
AK167900.1 | - | 4 | 667 | |||
AV446044.1 | - | 3 | 336 | |||
BC022974.1 | - | 4 | 647 | 5373335 | ||
M65255.1 | - | 4 | 644 |