Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnpat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010322.3 | NP_034452.3 | 14, 15 | 2142 | ||
GenBank mRNA | AF110769.1 | - | 14, 15 | 2106 | ||
AJ132012.1 | - | 14, 15 | 1937 | |||
AK004364.1 | - | 5, 6 | 589 | |||
AK004630.1 | - | 14, 15 | 2116 | |||
AK004771.1 | - | 14, 15 | 2122 | |||
AK010896.1 | - | 14, 15 | 2116 | |||
AK150810.1 | - | 14, 15 | 2100 | |||
AK166997.1 | - | 14, 15 | 2114 | |||
AK208037.1 | - | 4, 5 | 389 |