Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010350.2 | NP_034480.2 | 6 | 1805 | ||
GenBank mRNA | AK147390.1 | - | 5 | 1533 | ||
AK165770.1 | - | 3 | 660 | |||
BC137849.1 | - | 4 | 1195 | 8860868 | ||
D10694.1 | - | 3 | 1032 |