Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hemt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010416.2 | NP_034546.2 | 4 | 580 | ||
GenBank mRNA | AJ011022.1 | - | 2 | 503 | ||
AJ011023.1 | - | 2 | 465 | |||
AK005659.1 | - | 4 | 585 | |||
AK005823.1 | - | 3 | 353 | |||
BC119337.1 | - | 2 | 496 |
Gene Symbol: Gml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ242831.1 | - | 3 | 728 |