Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nr4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010444.2 | NP_034574.1 | 3 | 1066 | ||
GenBank mRNA | AK004709.1 | - | 3 | 1060 | ||
AK157203.1 | - | 3 | 1066 | |||
BC004770.1 | - | 3 | 1046 | 3156434 | ||
CT010287.1 | - | 2 | 938 | |||
J04113.1 | - | 3 | 1049 | |||
X60132.1 | - | 3 | 1016 |