Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RTN4 SIRNA Assays ›
Gene Symbol:
RTN4
Gene Name:
reticulon 4
Gene Aliases:
ASY, NI220/250, NOGO, NOGO-A, NOGOC, NSP, NSP-CL, Nbla00271, Nbla10545, Nogo-B, Nogo-C, RTN-X, RTN4-A, RTN4-B1, RTN4-B2, RTN4-C
Chromosome Location:
Chr.2: 54972187 - 55137831 on Build GRCh38
UniGene ID:
Hs.637850
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
6738
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RTN4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321859.1
NP_001308788.1
4
2727
NM_001321860.1
NP_001308789.1
4
2797
NM_001321861.1
NP_001308790.1
4
2760
NM_001321862.1
NP_001308791.1
4
2747
NM_001321863.1
NP_001308792.1
4
2697
NM_001321904.1
NP_001308833.1
5
3175
NM_007008.2
NP_008939.1
2
417
NM_020532.4
NP_065393.1
4
3480
NM_153828.2
NP_722550.1
2
1023
NM_207520.1
NP_997403.1
3
1080
NM_207521.1
NP_997404.1
4
2738
NR_135829.1
2
303
NR_135830.1
2
281
XM_005264434.3
XP_005264491.1
5
2874
XM_017004518.1
XP_016860007.1
5
2913
XM_017004519.1
XP_016860008.1
5
3030
GenBank mRNA
AB015639.1
-
2
791
AB020693.1
-
4
3316
AB040462.1
-
4
3426
AB040463.1
-
2
969
AB073351.1
-
2
1152
AF063601.1
-
2
1740
AF077050.1
-
2
416
AF087901.1
-
2
308
AF125103.1
-
2
417
AF132047.1
-
2
904
AF132048.1
-
2
323
AF148537.1
-
4
3323
AF148538.1
-
2
866
AF177332.1
-
2
322
AF320999.1
-
3
2486
AF333336.1
-
4
2742
AJ251383.1
-
4
3182
AJ251384.1
-
2
725
AJ251385.1
-
2
203
AK075039.1
-
2
957
AK129806.1
-
2
882
AK130812.1
-
2
784
AY102276.1
-
2
437
AY102277.1
-
2
969
AY102278.1
-
3
1026
AY102279.1
-
4
3426
AY123245.1
-
4
2739
AY123246.1
-
4
2797
AY123247.1
-
4
2760
AY123248.1
-
4
2746
AY123249.1
-
4
2707
AY123250.1
-
4
2697
BC001035.2
-
2
388
BC007109.1
-
2
314
BC010737.1
-
2
730
BC012619.1
-
2
880
BC014366.1
-
2
314
BC016165.1
-
3
1029
BC026788.1
-
2
911
BC068991.1
-
2
973
BC071848.1
-
3
711
BC150182.1
-
4
3430
40148210
BC152425.1
-
4
3316
8862539
BC152555.1
-
4
3430
40148984
CN429712.1
-
2
252
DQ778739.1
-
2
203
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_194054, NM_194053, NM_024226, NM_194051, NM_194052, and others.
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
axonal fasciculation
cerebral cortex radial glia guided migration
negative regulation of cell growth
negative regulation of axon extension
regulation of apoptotic process
negative regulation of axonogenesis
nuclear pore complex assembly
cardiac epithelial to mesenchymal transition
endoplasmic reticulum tubular network organization
endoplasmic reticulum tubular network assembly
cell-cell adhesion
regulation of branching morphogenesis of a nerve
protein binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7cf5v:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0