Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARD16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017534.1 | NP_001017534.1 | 3 | 363 | ||
NM_052889.2 | NP_443121.1 | 4 | 426 | |||
XM_011542583.2 | XP_011540885.1 | 3 | 555 | |||
GenBank mRNA | AF367017.1 | - | 3 | 400 | ||
AK125640.1 | - | 2 | 2947 | |||
AY885669.1 | - | 3 | 354 | |||
BC070196.1 | - | 4 | 426 | |||
BC104635.1 | - | 4 | 430 |