Gene Symbol: Itgb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145884.1 | NP_001139356.1 | 1, 2 | 374 | ||
NM_010580.2 | NP_034710.2 | 1, 2 | 374 | |||
GenBank mRNA | AF022110.1 | - | 1, 2 | 373 | ||
AF043256.1 | - | 1, 2 | 279 | |||
AF043257.1 | - | 1, 2 | 279 | |||
AK087476.1 | - | 1, 2 | 120 | |||
AK140251.1 | - | 1, 2 | 286 | |||
BC046280.1 | - | 1, 2 | 283 | 6394641 | ||
BC058246.1 | - | 1, 2 | 338 | 6313668 |