Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010601.3 | NP_034731.3 | 2 | 290 | ||
GenBank mRNA | AY380579.1 | - | 2 | 290 | ||
BC141013.1 | - | 2 | 405 | 9055625 | ||
BC145145.1 | - | 2 | 405 | 9053696 |