Gene Symbol: Itgb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145884.1 | NP_001139356.1 | 3 | 505 | ||
NM_010580.2 | NP_034710.2 | 3 | 505 | |||
GenBank mRNA | AF022110.1 | - | 3 | 504 | ||
AF043256.1 | - | 3 | 410 | |||
AF043257.1 | - | 3 | 410 | |||
AK087476.1 | - | 3 | 251 | |||
AK140251.1 | - | 3 | 417 | |||
AK143532.1 | - | 3 | 440 | |||
BC046280.1 | - | 3 | 414 | 6394641 | ||
BC058246.1 | - | 3 | 469 | 6313668 |