Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Jak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190830.1 | NP_001177759.1 | 3 | 446 | ||
NM_010589.6 | NP_034719.2 | 3 | 446 | |||
GenBank mRNA | AK038268.1 | - | 2 | 257 | ||
AK088365.1 | - | 3 | 298 | |||
AK142178.1 | - | 3 | 284 | |||
AK156646.1 | - | 3 | 268 | |||
AK164520.1 | - | 3 | 286 | |||
BC027234.1 | - | 277 | ||||
BC105577.1 | - | 2 | 249 | 3968386 | ||
BC131646.1 | - | 2 | 257 | 40110146 | ||
BC131647.1 | - | 2 | 257 | 40110148 | ||
L32955.1 | - | 2 | 248 | |||
L33768.1 | - | 2 | 242 | |||
L40172.1 | - | 3 | 307 |