Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010646.1 | NP_034776.1 | 5 | 635 | ||
GenBank mRNA | AF204265.1 | - | 6 | 626 | ||
AF204266.1 | - | 5 | 587 | |||
AF204267.1 | - | 6 | 617 | |||
AF204268.1 | - | 5 | 578 | |||
AF307947.1 | - | 5 | 639 | |||
AF307948.1 | - | 5 | 635 |