Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Jak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190830.1 | NP_001177759.1 | 10, 11 | 1649 | ||
NM_010589.6 | NP_034719.2 | 10, 11 | 1649 | |||
GenBank mRNA | AK038268.1 | - | 9, 10 | 1460 | ||
AK088365.1 | - | 10, 11 | 1498 | |||
AK142178.1 | - | 10, 11 | 1487 | |||
AK156646.1 | - | 10, 11 | 1471 | |||
AK164520.1 | - | 10, 11 | 1489 | |||
BC105577.1 | - | 9, 10 | 1452 | 3968386 | ||
BC131646.1 | - | 9, 10 | 1460 | 40110146 | ||
BC131647.1 | - | 9, 10 | 1460 | 40110148 | ||
L32955.1 | - | 9, 10 | 1448 | |||
L33768.1 | - | 7, 8 | 2093 | |||
L40172.1 | - | 10, 11 | 1510 |