Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010663.2 | NP_034793.1 | 4 | 839 | ||
GenBank mRNA | AB013608.1 | - | 4 | 840 | ||
BC032161.1 | - | 4 | 886 | |||
BC132454.1 | - | 4 | 874 | 40130731 | ||
BC132456.1 | - | 4 | 874 | 40130733 | ||
M13805.1 | - | 1 | 131 |