Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mecp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081979.2 | NP_001075448.1 | 2 | 271 | ||
NM_010788.4 | NP_034918.1 | 3 | 395 | |||
GenBank mRNA | AB221662.1 | - | 2 | 116 | ||
AF072251.1 | - | 2 | 116 | |||
AF158181.1 | - | 3 | 298 | |||
AJ132922.1 | - | 2 | 151 | |||
AK158664.1 | - | 2 | 218 | |||
BC027153.1 | - | 3 | 317 | 4948925 |