Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271416.1 | NP_001258345.1 | 391 | |||
NM_001271417.1 | NP_001258346.1 | 376 | ||||
NM_001271418.1 | NP_001258347.1 | 355 | ||||
NM_001271419.1 | NP_001258348.1 | 345 | ||||
NM_001271446.1 | NP_001258375.1 | 371 | ||||
NM_010738.3 | NP_034868.1 | 360 | ||||
GenBank mRNA | BC002070.1 | - | 2, 3 | 217 | 3486025 | |
BG085582.2 | - | 214 | ||||
BY228083.1 | - | 234 | ||||
EU704682.1 | - | 2, 3 | 205 | |||
J03636.1 | - | 224 | ||||
M18184.1 | - | 2, 3 | 191 |