Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010764.2 | NP_034894.2 | 8, 9 | 1164 | ||
GenBank mRNA | AK004817.2 | - | 8, 9 | 1121 | ||
AK035551.1 | - | 8, 9 | 1119 | |||
AK147928.1 | - | 8, 9 | 1122 | |||
AK150154.1 | - | 5, 6 | 496 | |||
AK170848.1 | - | 5, 6 | 495 | |||
AK171166.1 | - | 8, 9 | 1122 | |||
AK171377.1 | - | 5, 6 | 495 | |||
BC005430.1 | - | 8, 9 | 1109 | 3488681 | ||
U29947.1 | - | 8, 9 | 1107 | |||
U87240.1 | - | 8, 9 | 1098 |