Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mllt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252560.1 | NP_001239489.1 | 6 | 718 | ||
NM_001252561.1 | NP_001239490.1 | 7 | 769 | |||
NM_010804.4 | NP_034934.2 | 6 | 573 | |||
GenBank mRNA | AF010135.1 | - | 6 | 616 | ||
AK045042.1 | - | 6 | 473 | |||
AK147388.1 | - | 9 | 1063 | |||
AK154320.1 | - | 6 | 718 | |||
AK158426.1 | - | 5 | 311 | |||
AK220504.1 | - | 8 | 825 | |||
BC055298.1 | - | 7 | 718 | |||
BC069988.1 | - | 5 | 567 | |||
BC070475.1 | - | 6 | 539 | 30638433 | ||
BY608946.1 | - | 1 | 17 | |||
CN537566.1 | - | 3 | 246 |