Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010764.2 | NP_034894.2 | 11 | 1430 | ||
GenBank mRNA | AK004817.2 | - | 11 | 1387 | ||
AK035551.1 | - | 11 | 1385 | |||
AK147928.1 | - | 11 | 1388 | |||
AK150154.1 | - | 8 | 762 | |||
AK170848.1 | - | 8 | 761 | |||
AK171166.1 | - | 11 | 1388 | |||
AK171377.1 | - | 8 | 761 | |||
BC005430.1 | - | 11 | 1375 | 3488681 | ||
BC023332.1 | - | 2 | 132 | |||
U29947.1 | - | 11 | 1373 | |||
U87240.1 | - | 11 | 1364 |