Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC101943.1 | - | 5 | 748 | ||
BC103513.1 | - | 5 | 724 | |||
BC103514.1 | - | 5 | 748 | |||
BC103515.1 | - | 5 | 748 | |||
BC146612.1 | - | 5 | 720 | |||
BC146613.1 | - | 5 | 725 | 40039147 | ||
X73677.1 | - | 5 | 735 | |||
Z22649.1 | - | 5 | 725 |