Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010861.3 | NP_034991.3 | 2, 3 | 136 | ||
GenBank mRNA | AK002367.1 | - | 2, 3 | 137 | ||
AK146674.1 | - | 2, 3 | 140 | |||
BC061144.1 | - | 137 | 30301284 | |||
M91602.1 | - | 2, 3 | 128 |