Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfix | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081981.2 | NP_001075450.1 | 2 | 1264 | ||
NM_001081982.2 | NP_001075451.1 | 2 | 589 | |||
NM_001297601.1 | NP_001284530.1 | 2 | 1264 | |||
NM_010906.3 | NP_035036.1 | 2 | 589 | |||
GenBank mRNA | AK158568.1 | - | 2 | 1267 | ||
BC003766.1 | - | 2 | 311 | 3491917 | ||
BC090626.1 | - | 2 | 291 | |||
S81451.1 | - | 1 | 275 | |||
U57636.1 | - | 1 | 266 | |||
Y07688.1 | - | 2 | 591 | |||
Y07689.1 | - | 2 | 393 |