Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010907.2 | NP_035037.2 | 1 | 243 | ||
GenBank mRNA | AK089215.1 | - | 1 | 192 | ||
AK151115.1 | - | 1 | 196 | |||
AK151608.1 | - | 1 | 196 | |||
AK161622.1 | - | 1 | 154 | |||
BC046754.1 | - | 1 | 176 | 5693175 | ||
U36277.1 | - | 1 | 187 |